Anti-PASK Mouse Monoclonal Antibody [clone: 11D8]

GTX83940
89272-678EA 450.84 USD
89272-678
Anti-PASK Mouse Monoclonal Antibody [clone: 11D8]
Mouse monoclonal antibody [11D8] to PASK

Recommended Starting Dilutions:WB 1:2000 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PASK
Clonality: Monoclonal
Clone: 11D8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR