Anti-PBLD Mouse Monoclonal Antibody [clone: 4F6]

GTX83937
89272-672EA 450.84 USD
89272-672
Anti-PBLD Mouse Monoclonal Antibody [clone: 4F6]
Mouse monoclonal antibody [4F6] to PBLD

Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PBLD
Clonality: Monoclonal
Clone: 4F6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR