Anti-PDE10A Mouse Monoclonal Antibody [clone: 1B2]

Supplier: GeneTex
GTX83931
89272-662EA 705.72 USD
89272-662
Anti-PDE10A Mouse Monoclonal Antibody [clone: 1B2]
Antibodies
Mouse monoclonal antibody [1B2] to PDE10A

Recommended Starting Dilutions:ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PDE10A
Clonality: Monoclonal
Clone: 1B2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR