Anti-PECAM1 Mouse Monoclonal Antibody [clone: 8A9]

GTX83915
89272-630EA 571.49 USD
89272-630
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 8A9]
Mouse monoclonal antibody [8A9] to CD31

Recommended Starting Dilutions:Western blot: 1:500 IHC 1:50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 8A9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR