Anti-PECR Mouse Monoclonal Antibody [clone: 10000]

Supplier: GeneTex
GTX83911
89272-622EA 705.1 USD
89272-622
Anti-PECR Mouse Monoclonal Antibody [clone: 10000]
Antibodies
Mouse monoclonal antibody [1E4] to PECR

Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PECR
Clonality: Monoclonal
Clone: 10000
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR