Anti-PEPD Mouse Monoclonal Antibody [clone: 1B7]

Supplier: GeneTex
GTX83909
89272-618EA 705.34 USD
89272-618
Anti-PEPD Mouse Monoclonal Antibody [clone: 1B7]
Antibodies
Mouse monoclonal antibody [1B7] to PEPD

Recommended Starting Dilutions: WB 1:2000 IHC 1:150 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PEPD
Clonality: Monoclonal
Clone: 1B7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR