Anti-Troponin I Type 3 Mouse Monoclonal Antibody [clone: 4A3]

Supplier: GeneTex
GTX83501
89271-810EA 705.1 USD
89271-810
Anti-Troponin I Type 3 Mouse Monoclonal Antibody [clone: 4A3]
Antibodies
Mouse monoclonal antibody [4A3] to Cardiac Troponin I

Recommended Starting Dilutions:WB 1:2000 IHC 1:50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: Troponin I Type 3
Clonality: Monoclonal
Clone: 4A3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR