Anti-HK1 Mouse Monoclonal Antibody [clone: 7A7]

Supplier: GeneTex
GTX82795
89287-244EA 618.44 USD
89287-244
Anti-HK1 Mouse Monoclonal Antibody [clone: 7A7]
Antibodies
Mouse monoclonal antibody [7A7] to HK1

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.IHC: 1/200 - 1/1000.ICC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: HK1
Clonality: Monoclonal
Clone: 7A7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR