Anti-LAG3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81605
89302-296EA 683.58 USD
89302-296
Anti-LAG3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to LAG3 (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: LAG3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR