Anti-AMHR2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81590
89302-266EA 584.29 USD
89302-266
Anti-AMHR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to AMHR2 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: AMHR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR