Anti-INS (Insulin) (insulin) Rabbit Polyclonal Antibody

GTX81555
89302-196EA 403.5 USD
89302-196
Anti-INS (Insulin) (insulin) Rabbit Polyclonal Antibody
Rabbit polyclonal to INS (Insulin) (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: INS (Insulin) (insulin)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR