Anti-HNF4G Rabbit Polyclonal Antibody

GTX81438
89301-962EA 403.86 USD
89301-962
Anti-HNF4G Rabbit Polyclonal Antibody
Rabbit polyclonal to HNF4G (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: HNF4G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR