Anti-CD31 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81432
89301-950EA 610.6 USD
89301-950
Anti-CD31 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CD31 (C-term)

Recommended Starting Dilutions: WB: Use at 1:1000, IHC-P: Use at 1:10-50, FACS: Use at 1:10-50. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CD31
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR