Anti-EPHA6 Rabbit Polyclonal Antibody

GTX81359
89301-808EA 403.86 USD
89301-808
Anti-EPHA6 Rabbit Polyclonal Antibody
Rabbit polyclonal to EphA6

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: EPHA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR