Anti-NTF3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81308
89301-706EA 610.6 USD
89301-706
Anti-NTF3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to NTF3 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: NTF3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR