Anti-CTGF Rabbit Polyclonal Antibody

GTX81298
89301-686EA 403.86 USD
89301-686
Anti-CTGF Rabbit Polyclonal Antibody
Rabbit polyclonal to CTGF (C-term)

  • Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100IHC: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR