Anti-CCNC Rabbit Polyclonal Antibody

GTX81283
89301-656EA 403.86 USD
89301-656
Anti-CCNC Rabbit Polyclonal Antibody
Rabbit polyclonal to Cyclin C (CCNC) (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CCNC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR