Anti-FKBP1B Rabbit Polyclonal Antibody

GTX81258
89301-606EA 403.86 USD
89301-606
Anti-FKBP1B Rabbit Polyclonal Antibody
Rabbit polyclonal to FKBP1B (N-term E6)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: FKBP1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR