Anti-SLC11A1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81205
89301-502EA 610.43 USD
89301-502
Anti-SLC11A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SLC11A1 (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: SLC11A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR