Anti-MPZ Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81135
89301-364EA 610.91 USD
89301-364
Anti-MPZ Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to MPZ (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500FACS: Use at 1:10-50IHC: Use at 1:50~100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: MPZ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR