Anti-TARSL2 (threonyl-tRNA Synthetase-like 2) Rabbit Polyclonal Antibody

GTX81116
89301-326EA 403.86 USD
89301-326
Anti-TARSL2 (threonyl-tRNA Synthetase-like 2) Rabbit Polyclonal Antibody
Rabbit polyclonal to TARSL2 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: TARSL2 (threonyl-tRNA Synthetase-like 2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR