Anti-FLCN (folliculin) Rabbit Polyclonal Antibody

GTX81112
89301-318EA 403.86 USD
89301-318
Anti-FLCN (folliculin) Rabbit Polyclonal Antibody
Rabbit polyclonal to FLCN (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: FLCN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR