Anti-KMO Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81111
89301-316EA 610.91 USD
89301-316
Anti-KMO Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to KMO (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: KMO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR