Anti-ATF6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81096
89301-288EA 610.6 USD
89301-288
Anti-ATF6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to ATF6 (Internal)

  • Recommended Starting Dilutions: For WB: Use at a dilution of 1:1000. For IHC-P: Use at a dilution of 1:10-50. For FACS: Use at a dilution of 1:10-50. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: ATF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR