Anti-C10ORF27 Rabbit Polyclonal Antibody

GTX81090
89301-276EA 403.86 USD
89301-276
Anti-C10ORF27 Rabbit Polyclonal Antibody
Rabbit polyclonal to C10orf27 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: C10ORF27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR