Anti-CACNA2D1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81084
89301-264EA 610.6 USD
89301-264
Anti-CACNA2D1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cav alpha 2

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CACNA2D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR