Anti-AGPAT4 Rabbit Polyclonal Antibody

GTX81079
89301-254EA 403.86 USD
89301-254
Anti-AGPAT4 Rabbit Polyclonal Antibody
Rabbit polyclonal to AGPAT4 (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: AGPAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR