Anti-CDC42EP3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81035
89301-168EA 610.6 USD
89301-168
Anti-CDC42EP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CDC42EP3 (N-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CDC42EP3
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR