Anti-TAZ Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX81015
89301-130EA 610.6 USD
89301-130
Anti-TAZ Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to TAZ (N-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: TAZ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR