Anti-SLC8A1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX80928
89300-960EA 610.6 USD
89300-960
Anti-SLC8A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SLC8A1 (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: SLC8A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR