Anti-IL1F8 Rabbit Polyclonal Antibody

GTX80917
89300-938EA 403.86 USD
89300-938
Anti-IL1F8 Rabbit Polyclonal Antibody
Rabbit polyclonal to IL1F8 (N-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: IL1F8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR