Anti-EMX1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX80913
89300-930EA 610.91 USD
89300-930
Anti-EMX1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to EMX1 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500Flow cytometric at 1:10~50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: EMX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR