Anti-Marburg virus Mouse Monoclonal Antibody [clone: 3545]

Supplier: GeneTex
GTX80771
89289-262EA 442.71 USD
89289-262
Anti-Marburg virus Mouse Monoclonal Antibody [clone: 3545]
Type: Primary
Antigen: Marburg virus
Clonality: Monoclonal
Clone: 3545
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR