Anti-Influenza B Mouse Monoclonal Antibody [clone: 1131]

Supplier: GeneTex
GTX80759
89289-254EA 579.08 USD
89289-254
Anti-Influenza B Mouse Monoclonal Antibody [clone: 1131]
Antibodies
Type: Primary
Antigen: Influenza B
Clonality: Monoclonal
Clone: 1131
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR