Anti-CPON Guinea Pig Polyclonal Antibody

Supplier: GeneTex
GTX80749
89285-556EA 379.61 USD
89285-556
Anti-CPON Guinea Pig Polyclonal Antibody
Type: Primary
Antigen: CPON
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea Pig
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR