Anti-NACC1 Mouse Monoclonal Antibody [clone: 6H2]

Supplier: GeneTex
GTX80403
89291-684EA 618.44 USD
89291-684
Anti-NACC1 Mouse Monoclonal Antibody [clone: 6H2]
Antibodies
Mouse Monoclonal antibody [6H2] to NACC1

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.IHC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: NACC1
Clonality: Monoclonal
Clone: 6H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR