Anti-FCER2 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX80402
89291-682EA 618.44 USD
89291-682
Anti-FCER2 Mouse Monoclonal Antibody
Antibodies
Mouse Monoclonal antibody to FCER2

Recommended Starting Dilutions:ELISA: 1/10000.FCM: 1/200 - 1/400.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: FCER2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR