Anti-CTCF (CCCTC-binding Factor (zinc finger Protein)) Mouse Monoclonal Antibody [clone: 1D11]

Supplier: GeneTex
GTX80397
89291-674EA 618.44 USD
89291-674
Anti-CTCF (CCCTC-binding Factor (zinc finger Protein)) Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Mouse Monoclonal antibody [1D11] to CTCF

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: CTCF (CCCTC-binding Factor (zinc finger Protein))
Clonality: Monoclonal
Clone: 1D11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR