Anti-HIV-2 Nef Rat Monoclonal Antibody [clone: 4B5 ]

Supplier: GeneTex
GTX80392
89276-240EA 738.97 USD
89276-240
Anti-HIV-2 Nef Rat Monoclonal Antibody [clone: 4B5 ]
Antibodies
Rat Monoclonal antibody [4B5] to HIV-2 Nef

Recommended Starting Dilutions: For WB: Use at 1:1000. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: HIV-2 Nef
Clonality: Monoclonal
Clone: 4B5 
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 
Reactivity: HIV
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR