Anti-HNRNPL Mouse Monoclonal Antibody [clone: 4D11 ]

Supplier: GeneTex
GTX80386
89300-454EA 480.68 USD
89300-454
Anti-HNRNPL Mouse Monoclonal Antibody [clone: 4D11 ]
Antibodies
Mouse Monoclonal antibody [4D11] to hnRNP-L

Recommended Starting Dilutions:For WB: Use at 1:5000Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: HNRNPL
Clonality: Monoclonal
Clone: 4D11 
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Newt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR