Anti-ORC6 Rat Monoclonal Antibody [clone: 3A4 ]

Supplier: GeneTex
GTX80357
89280-880EA 748.01 USD
89280-880
Anti-ORC6 Rat Monoclonal Antibody [clone: 3A4 ]
Antibodies
Rat Monoclonal antibody [3A4] to ORC6

Recommended Starting Dilutions:For WB: Use at 1:1000Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: ORC6
Clonality: Monoclonal
Clone: 3A4 
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR