Anti-DPH1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX80356
89300-422EA 683.14 USD
89300-422
Anti-DPH1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to OVCA1

Recommended Starting Dilutions:For WB: Use at 1:1000Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: DPH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR