Anti-PGR Mouse Monoclonal Antibody [clone: 1064/E2]

Supplier: GeneTex
GTX80351
89300-414EA 606.4 USD
89300-414
Anti-PGR Mouse Monoclonal Antibody [clone: 1064/E2]
Antibodies
Mouse Monoclonal antibody [1064/E2] to Progesterone Receptor (phospho Ser162)

Recommended Starting Dilutions:Use at 5-10µg/mlOptimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 1064/E2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR