Anti-Pseudomonas aeruginosa serotype 6c Mouse Monoclonal Antibody [clone: MJ6302 ]

Supplier: GeneTex
GTX80346
89300-404EA 606.4 USD
89300-404
Anti-Pseudomonas aeruginosa serotype 6c Mouse Monoclonal Antibody [clone: MJ6302 ]
Antibodies
Type: Primary
Antigen: Pseudomonas aeruginosa serotype 6c
Clonality: Monoclonal
Clone: MJ6302 
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR