Anti-TAP1 Mouse Monoclonal Antibody [clone: 53H8 ]

Supplier: GeneTex
GTX80332
89300-380EA 606.4 USD
89300-380
Anti-TAP1 Mouse Monoclonal Antibody [clone: 53H8 ]
Antibodies
Mouse Monoclonal antibody [53H8] to TAP

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: TAP1
Clonality: Monoclonal
Clone: 53H8 
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR