Anti-Trichohyalin granules (THG) Mouse Monoclonal Antibody [clone: AE15 ]

Supplier: GeneTex
GTX80330
89300-378EA 618.66 USD
89300-378
Anti-Trichohyalin granules (THG) Mouse Monoclonal Antibody [clone: AE15 ]
Antibodies
Type: Primary
Antigen: Trichohyalin granules (THG)
Clonality: Monoclonal
Clone: AE15 
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR