Anti-Glu-Glu tag Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77387
89317-836EA 628.47 USD
89317-836
Anti-Glu-Glu tag Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Glu-Glu tag
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR