Anti-CD229 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HLy 9.1.25]

Supplier: GeneTex
GTX75856
89265-608EA 507.51 USD
89265-608
Anti-CD229 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HLy 9.1.25]
Antibodies
Mouse monoclonal antibody [HLy9.1.25] to CD229

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: HLy 9.1.25
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR