Anti-CD229 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HLy 9.1.25]

GTX75855
89279-940EA 442.18 USD
89279-940
Anti-CD229 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HLy 9.1.25]
Mouse monoclonal antibody [HLy9.1.25] to CD229

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: HLy 9.1.25
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR