Anti-CD229 Mouse Monoclonal Antibody [clone: HLy 9.1.25]

Supplier: GeneTex
GTX75853
89280-814EA 755.08 USD
89280-814
Anti-CD229 Mouse Monoclonal Antibody [clone: HLy 9.1.25]
Antibodies
Mouse monoclonal antibody [HLy9.1.25] to CD229

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: HLy 9.1.25
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR